Zinc formate dihydrate ≥98%

Supplier: BeanTown Chemical

216775-1KG 216775-250G
BT216775-1KGEA 114 USD
BT216775-1KG BT216775-250G
Zinc formate dihydrate ≥98%
Zinc formate dihydrate

CAS: 5970-62-7; EC No: 209-176-5; MDL No: MFCD00050836 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: Zn(CHO2)2·2H2O; MW: 191.45 Density (g/mL): 2.201


  • Powder

Formula: C₂H₆O₆Zn
Density: 2.201 g/mL
CAS Number: 5970-62-7
EINECS: 209-176-5
ADR: 9,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR