α,4-Dichlorotoluene ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Chloro-4-(chloromethyl)benzene, 4-Chlorobenzyl chloride

216745-1KG 216745-250G
BT216745-1KGEA 159 USD
BT216745-1KG BT216745-250G
α,4-Dichlorotoluene ≥98%
α,4-Dichlorotoluene

CAS: 104-83-6; EC No: 203-242-7; MDL No: MFCD00000914; RTECS: XT0720000 UN No: UN3427; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C7H6Cl2; MW: 161.03 Melting Point: 27-29°; Boiling Point: 216-222°; Flash point: 108°C (226°F) Density (g/mL): 1.275


  • RTECS: XT0720000
  • Solid

Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 216…222 °C
Melting Pt: 27…29 °C
Density: 1.275 g/mL
Flash Pt: 108 °C (226 °F)
MDL Number: MFCD00000914
CAS Number: 104-83-6
EINECS: 203-242-7
UN: 3427
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR