4-Iodo-o-toluidine ≥98%

Supplier: BeanTown Chemical

216695-25G 216695-5G
BT216695-25GEA 268 USD
BT216695-25G BT216695-5G
4-Iodo-o-toluidine ≥98%
4-Iodo-o-toluidine

CAS: 13194-68-8; EC No: 236-154-2; MDL No: MFCD00025299 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C7H8IN ; MW: 233.05 Melting Point: 86-89° Light Sensitive


  • Light Sensitive
  • Powder

Formula: C₇H₈IN
Melting Pt: 86…89 °C
MDL Number: MFCD00025299
CAS Number: 13194-68-8
EINECS: 236-154-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR