N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7

216675-1G 216675-5G 216675-5X1G
BT216675-1GEA 123.33 USD
BT216675-1G BT216675-5G BT216675-5X1G
N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic)
N,N-Dimethylformamide-[D₇]

CAS: 4472-41-7; EC No: 224-745-8; MDL No: MFCD00003286 UN No: UN2265; Haz Class: 3; Packing Group: III Liquid; Linear Formula: DCON(CD3)2; Molecular Formula: C3D7NO; MW: 80.14 Boiling Point: 153°; Flash point: 58°C (136°F) Density (g/mL): 1.03; Refractive Index: 1.428 Hygroscopic


  • Refractive index: 1.428
  • Hygroscopic
  • Liquid

Formula: DCOON(CD₃)₂
MW: 80.04 g/mol
Boiling Pt: 152
Melting Pt: -60
Density: 1.03 g/mL
Flash Pt: 58 °C (136 °F)
MDL Number: MFCD00003286
CAS Number: 4472-41-7
EINECS: 224-745-8
UN: 2265
ADR: 3 III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR