Butyl acrylate ≥98% stabilized

Supplier: BeanTown Chemical

Synonyms: n-Butyl acrylate, Acrylic acid butyl ester

216660-100ML 216660-2.5L 216660-500ML
BT216660-100MLEA 30.17 USD
BT216660-100ML BT216660-2.5L BT216660-500ML
Butyl acrylate ≥98% stabilized
Butyl acrylate

CAS: 141-32-2; EC No: 216-768-7; MDL No: MFCD00009446; RTECS: UD3150000 UN No: UN2348; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH2=CHCOO(CH2)3CH3; Molecular Formula: C7H12O2; MW: 128.17 Melting Point: -69°; Boiling Point: 145°; Flash point: 36°C (97°F) Density (g/mL): 0.894; Refractive Index: 1.418 Light Sensitive


  • RTECS: UD3150000
  • Refractive index: 1.418
  • Light Sensitive
  • Liquid

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 145 °C
Melting Pt: –69 °C
Density: 0.894 g/mL
Flash Pt: 36 °C (97 °F)
MDL Number: MFCD00008809
CAS Number: 141-32-2
EINECS: 216-768-7
UN: 2348
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR