Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol
Supplier: BeanTown Chemical
Synonyms:
Zirconium(IV) Tetrapropoxide, Zirconium(IV) n-propoxide
216655-2KG
216655-500G
216655-50G
BT216655-2KGEA
391
USD
BT216655-2KG
BT216655-500G
BT216655-50G
Zirconium(IV) Propoxide ≥70% (w/w) in n-propanol
Zirconium(IV) Propoxide
CAS: 23519-77-9; EC No: 245-711-9; MDL No: MFCD00015307
UN No: UN1274; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: Zr(OCH2CH2CH3)4; Molecular Formula: C12H28O4Zr; MW: 327.57
Boiling Point: 208°/0.1 mmHg; Flash point: 28°C (83°F)
Density (g/mL): 1.044; Refractive Index: 1.451
Moisture Sensitive
- Refractive index: 1.451
- Moisture Sensitive
- Liquid
Formula:
C₁₂H₂₈O₄Zr MW: 327.58 g/mol Boiling Pt: 208 °C/0.1 mmHg Density: 1.044 g/mL Flash Pt: 28 °C (83 °F) |
MDL Number:
MFCD00015307 CAS Number: 23519-77-9 EINECS: 245-711-9 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...