Trimethyl orthovalerate ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,1,1-Trimethoxypentane, Orthovaleric Acid Trimethyl Ester
216575-100G
216575-25G
216575-5G
BT216575-100GEA
375
USD
BT216575-100G
BT216575-25G
BT216575-5G
Trimethyl orthovalerate ≥98%
Trimethyl orthovalerate
CAS: 13820-09-2; EC No: 237-496-5; MDL No: MFCD00008481
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3CH2CH2CH2C(OCH3)3; Molecular Formula: C8H18O3; MW: 162.23
Boiling Point: 164-166°; Flash point: 41°C (105°F)
Density (g/mL): 0.941; Refractive Index: 1.410
Moisture Sensitive
- Refractive index: 1.410
- Moisture Sensitive
- Liquid
Formula:
C₈H₁₈O₃ MW: 162.23 g/mol Boiling Pt: 164…166 °C Density: 0.941 g/mL Flash Pt: 41 °C (105 °F) |
MDL Number:
MFCD00008481 CAS Number: 13820-09-2 EINECS: 237-496-5 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...