Rhodium ≥99.95% (trace metals basis), Powder -325 mesh
Supplier: BeanTown Chemical
216555-2G
216555-500MG
BT216555-2GEA
2681
USD
BT216555-2G
BT216555-500MG
Rhodium ≥99.95% (trace metals basis), Powder -325 mesh
Rhodium
CAS: 7440-16-6; EC No: 231-125-0; MDL No: MFCD00011201; RTECS: VI9069000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Rh; MW: 102.91
Melting Point: 1966°; Boiling Point: 3727°
Density (g/mL): 12.41
- RTECS: VI9069000
- Powder
Formula:
Rh MW: 102.9 g/mol Boiling Pt: 3727 °C Melting Pt: 1966 °C Density: 12.41 g/mL |
MDL Number:
MFCD00011201 CAS Number: 7440-16-6 EINECS: 231-125-0 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.95% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...