ω,4-Dibromoacetophenone ≥98%

Supplier: BeanTown Chemical

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

216505-100G 216505-25G 216505-5G
BT216505-100GEA 235 USD
BT216505-100G BT216505-25G BT216505-5G
ω,4-Dibromoacetophenone ≥98%
ω,4-Dibromoacetophenone

CAS: 99-73-0; EC No: 202-783-6; MDL No: MFCD00000200; RTECS: AM6950000 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C8H6Br2O; MW: 277.94 Melting Point: 108-110°


  • RTECS: AM6950000
  • Crystalline

Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Melting Pt: 108…110 °C
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR