Silver methanesulfonate

Supplier: BeanTown Chemical

216500-10G 216500-50G
BT216500-10GEA 120 USD
BT216500-10G BT216500-50G
Silver methanesulfonate
Silver methanesulphonate

CAS: 2386-52-9; EC No: 219-199-2; MDL No: MFCD00064795; RTECS: PB2685000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Linear Formula: AgSO3CH3; MW: 202.97 Melting Point: 252-256° Light Sensitive


  • RTECS: PB2685000
  • Light Sensitive
  • Powder

Formula: CH₃AgO₃S
MW: 202.97 g/mol
Melting Pt: 252…256 °C
MDL Number: MFCD00064795
CAS Number: 2386-52-9
EINECS: 219-199-2
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR