Dichloromethane-[D₂] ≥99.9% (Isotopic)
Supplier: BeanTown Chemical
216480-10EACH
216480-2EACH
BT216480-10EAEA
232.79
USD
BT216480-10EA
BT216480-2EACH
Dichloromethane-[D₂] ≥99.9% (Isotopic)
Dichloromethane-[D₂]
CAS: 1665-00-5; EC No: 216-776-0; MDL No: MFCD00000882
UN No: UN1593; Haz Class: 6.1; Packing Group: III
Liquid, in prescored ampules, 0.75ml/ampule ; Molecular Formula: CD2Cl2; MW: 86.95
Boiling Point: 40°
Density (g/mL): 1.362; Refractive Index: 1.422
Moisture Sensitive
- Refractive index: 1.422
- Moisture Sensitive
- Liquid, in prescored ampules, 0.75ml/ampule
Formula:
CD₂Cl₂ Boiling Pt: 40 °C Density: 1.362 g/mL |
MDL Number:
MFCD00000882 CAS Number: 1665-00-5 EINECS: 216-776-0 UN: 1593 ADR: 6.1,III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...