1-(4-Aminophenyl)-4-methylpiperazine ≥97%
Supplier: BeanTown Chemical
216435-1G
216435-5G
BT216435-1GEA
207
USD
BT216435-1G
BT216435-5G
1-(4-Aminophenyl)-4-methylpiperazine ≥97%
1-(4-Aminophenyl)-4-methylpiperazine
CAS: 16153-81-4; MDL No: MFCD00172703
UN No: UN3263; Haz Class: 8; Packing Group: III
Crystalline/Powder; Molecular Formula: C11H17N3; MW: 191.27
Melting Point: 89-95°
Air Sensitive
- Air Sensitive
- Crystalline/Powder
Formula:
C₁₁H₁₇N₃ Melting Pt: 89…95 °C |
MDL Number:
MFCD00172703 CAS Number: 16153-81-4 UN: 3263 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...