Trilithium nitride ≥99% (trace metals basis)

Supplier: BeanTown Chemical

216310-10G 216310-50G
BT216310-10GEA 62.97 USD
BT216310-10G BT216310-50G
Trilithium nitride ≥99% (trace metals basis)
tri-Lithium nitride

CAS: 26134-62-3; EC No: 247-475-2; MDL No: MFCD00016186 UN No: UN2806; Haz Class: 4.3; Packing Group: I -60 Mesh Powder; Molecular Formula: Li3N ; MW: 34.82 Melting Point: 840-850° Density (g/mL): 1.3 Moisture Sensitive


  • Moisture Sensitive
  • -60 Mesh Powder

Formula: Li₃N
MW: 34.83 g/mol
Melting Pt: 840…850 °C
Density: 1.3 g/mL
MDL Number: MFCD00016186
CAS Number: 26134-62-3
EINECS: 247-475-2
UN: 2806
ADR: 4.3,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR