Acetone ≥99.5% ACS
Supplier: BeanTown Chemical
CAS: 67-64-1; EC No: 200-662-2; MDL No: MFCD00008765; RTECS: AL3150000
UN No: UN1090; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3COCH3; MW: 58.08
Melting Point: -94°; Boiling Point: 56°; Flash point: -17°C (1°F)
Density (g/mL): 0.791; Refractive Index: 1.359
- RTECS: AL3150000
- Refractive index: 1.359
- Liquid
Formula:
CH₃COCH₃ Boiling Pt: 56 °C Melting Pt: –94 °C Density: 0.791 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00008765 CAS Number: 67-64-1 EINECS: 200-662-2 UN: 1090 ADR: 3,II |
|
Specification Test Results
Assay | 99.5% min. |
Specific gravity at 25°C | ≤0.7890 |
Color (APHA) | ≤10 |
Residue after evaporation | ≤10ppm |
Non-volatile residue | ≤0.004% |
Titrable acid | ≤0.003meq/g |
Titrable base | ≤0.006meq/g |
Aldehyde (as HCHO) | ≤0.002% |
IPA | ≤0.05% |
Methanol | ≤0.05% |
Specific Gravity 60/60F | 0.869-0.873 |
Water | ≤0.5% |
Identification, IR | Conforms to spectra |
Identification, GC | Conforms to chromatogram |
Appearance | Clear liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...