4,4,4-Trifluorobutyric acid ≥97%
Supplier: BeanTown Chemical
216225-1G
216225-5G
BT216225-1GEA
86
USD
BT216225-1G
BT216225-5G
4,4,4-Trifluorobutyric acid ≥97%
4,4,4-Trifluorobutyric acid
CAS: 406-93-9; MDL No: MFCD00077604
UN No: UN3261; Haz Class: 8; Packing Group: II
Low Melting Solid; Linear Formula: CF3CH2CH2CO2H; Molecular Formula: C4H5F3O2; MW: 142.08
Melting Point: 25-31°
Refractive Index: 1.351
- Refractive index: 1.351
- Low Melting Solid
Formula:
C₄H₅F₃O₂ MW: 142.08 g/mol Melting Pt: 25…31 °C |
MDL Number:
MFCD00077604 CAS Number: 406-93-9 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...