trans-2,3-Epoxybutane ≥97%

Supplier: BeanTown Chemical

216210-1G 216210-25G 216210-5G
BT216210-1GEA 43 USD
BT216210-1G BT216210-25G BT216210-5G
trans-2,3-Epoxybutane ≥97%
trans-2,3-Epoxybutane

CAS: 21490-63-1; EC No: 244-406-8; MDL No: MFCD00005129; RTECS: EK3855040 UN No: UN3271; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H8O ; MW: 72.11 Boiling Point: 54-55°; Flash point: -26°C (-15°F) Density (g/mL): 0.804 ; Refractive Index: 1.373 Moisture Sensitive


  • RTECS: EK3855040
  • Refractive index: 1.373
  • Moisture Sensitive
  • Liquid

Formula: C₄H₈O
MW: 72.11
Boiling Pt: 54…55 °C
Density: 0.804 g/mL
Flash Pt: –26 °C (–15 °F)
MDL Number: MFCD00005129
CAS Number: 21490-63-1
EINECS: 244-406-8
UN: 3271
ADR: 3 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR