3-Methylpentane ≥99%
Supplier: BeanTown Chemical
216200-100ML
216200-500ML
BT216200-100MLEA
47.78
USD
BT216200-100ML
BT216200-500ML
3-Methylpentane ≥99%
3-Methylpentane
CAS: 96-14-0; EC No: 202-481-4; MDL No: MFCD00009342
UN No: UN1208; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3CH2)2CH2CH3; Molecular Formula: C6H14; MW: 86.18
Boiling Point: 64°; Flash point: -7°C (19°F)
Density (g/mL): 0.664; Refractive Index: 1.376
- Refractive index: 1.376
- Liquid
Formula:
C₆H₁₄ MW: 86.18 g/mol Boiling Pt: 64 °C Density: 0.664 g/mL Flash Pt: –7 °C (19 °F) |
MDL Number:
MFCD00009342 CAS Number: 96-14-0 EINECS: 202-481-4 UN: 1208 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...