Acetic acid glacial ≥99.998% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Vinegar acid, Methanecarboxylic acid, Ethanoic acid
216175-100ML
216175-25ML
216175-500ML
BT216175-100MLEA
102
USD
BT216175-100ML
BT216175-25ML
BT216175-500ML
Acetic acid glacial ≥99.998% (trace metals basis)
Acetic acid
CAS: 64-19-7; EC No: 200-580-7; MDL No: MFCD00036152
UN No: UN2789; Haz Class: 8 (3); Packing Group: II
Liquid; Linear Formula: CH3CO2H; MW: 60.05
Melting Point: 16.2°; Boiling Point: 117-118°; Flash point: 40°C (104°F)
Density (g/mL): 1.049; Refractive Index: 1.371
- Refractive index: 1.371
- Liquid
Formula:
H₃CCOOH MW: 60.05 g/mol Boiling Pt: 117…118 °C Melting Pt: 16.2 °C Density: 1.049 g/mL Flash Pt: 40 °C (104 °F) |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8,II |
Specification Test Results
Assay | 99.998% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...