Potassium hydroxide ≥85% ACS

Supplier: BeanTown Chemical

216145-2KG 216145-500G
BT216145-2KGEA 123.48 USD
BT216145-2KG BT216145-500G
Potassium hydroxide ≥85% ACS
Potassium hydroxide

CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553; RTECS: TT2100000 UN No: UN1813; Haz Class: 8; Packing Group: II Pellets; Linear Formula: KOH; MW: 56.11 Melting Point: 361°; Boiling Point: 1320° Density (g/mL): 2.044 Air Sensitive , Hygroscopic


  • RTECS: TT2100000
  • Air Sensitive, Hygroscopic
  • Pellets

Formula: KOH
Boiling Pt: 1320 °C
Melting Pt: 361 °C
Density: 2.044 g/mL
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Assay 85.0% min.
Potassium carbonate (K2CO3) ≤2.0%
Chloride (Cl) ≤0.01%
Nitrogen compounds (as N) ≤0.001%
Phosphate (PO4) ≤5ppm
Sulfate (SO4) ≤0.003%
Heavy metals (as Ag) ≤0.001%
Iron (Fe) ≤0.001%
Nickel (Ni) ≤0.001%
Calcium (Ca) ≤0.005%
Magnesium (Mg) ≤0.002%
Sodium (Na) ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR