Sodium triacetoxyborohydride ≥95%

Supplier: BeanTown Chemical

216085-100G 216085-25G 216085-5G
BT216085-100GEA 170.45 USD
BT216085-100G BT216085-25G BT216085-5G
Sodium triacetoxyborohydride ≥95%
Sodium triacetoxyborohydride

CAS: 56553-60-7; MDL No: MFCD00012211 UN No: UN3396; Haz Class: 4.3 (4.1); Packing Group: III Solid; Linear Formula: (CH3COO)3BHNa; MW: 211.94 Melting Point: 116-120° Density (g/mL): 1.43 Moisture Sensitive


  • Moisture Sensitive
  • Solid

Formula: C₆H₁₀BNaO₆
MW: 211.94 g/mol
Melting Pt: 116…120 °C
Density: 1.43 g/mL
MDL Number: MFCD00012211
CAS Number: 56553-60-7
UN: 3396
ADR: 4.3,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR