1-Bromo-4-fluoro-3-nitrobenzene ≥98%

Supplier: BeanTown Chemical

216080-1G 216080-25G 216080-5G
BT216080-1GEA 28 USD
BT216080-1G BT216080-25G BT216080-5G
1-Bromo-4-fluoro-3-nitrobenzene ≥98%
1-Bromo-4-fluoro-3-nitrobenzene

CAS: 364-73-8; MDL No: MFCD00129165 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid/Low Melting Solid; Molecular Formula: C6H3BrFNO2; MW: 220.00 Melting Point: 18-19°; Boiling Point: 240-241°; Flash point: 110°C (230°F) Density (g/mL): 1.786; Refractive Index: 1.575


  • Refractive index: 1.575
  • Liquid/Low Melting Solid

Formula: C₆H₃BrFNO₂
Boiling Pt: 240…241 °C
Melting Pt: 18…19 °C
Density: 1.786 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00129165
CAS Number: 364-73-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR