Ammonium metavanadate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Ammonium monovanadate, Ammonium vanadium trioxide, Ammonium vanadium oxide

215995-250G 215995-50G
BT215995-250GEA 141 USD
BT215995-250G BT215995-50G
Ammonium metavanadate ≥99.0% ACS
Ammonium metavanadate

CAS: 7803-55-6; EC No: 232-261-3; MDL No: MFCD00011430 UN No: UN2859; Haz Class: 6.1; Packing Group: II Powder; Linear Formula: NH4VO3; MW: 116.98 Density (g/mL): 2.32


  • Powder

Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200
Density: 2.32 g/mL
MDL Number: MFCD00011430
CAS Number: 7803-55-6
EINECS: 232-261-3
UN: 2859
ADR: 6.1 II
Merck Index: 13,00572

Order Now

Specification Test Results

Assay 99.0% min.
Solubility in NH4OH Passes test
Carbonate (CO3) Passes test
Chloride (Cl) ≤0.2%
Sulfate (SO4) ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR