Dimethylethoxysilane ≥94%

Supplier: BeanTown Chemical

215920-100G 215920-25G
BT215920-100GEA 197 USD
BT215920-100G BT215920-25G
Dimethylethoxysilane ≥94%
Dimethylethoxysilane

CAS: 14857-34-2; EC No: 238-921-7; MDL No: MFCD00026750; RTECS: VV4090000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4H12OSi; MW: 104.23 Boiling Point: 55°; Flash point: 15°C (59°F) Density (g/mL): 0.757 Moisture Sensitive


  • RTECS: VV4090000
  • Moisture Sensitive
  • Liquid

Formula: C₄H₁₂OSi
MW: 104.23 g/mol
Boiling Pt: 55 °C
Density: 0.757 g/mL
Flash Pt: 15 °C (59 °F)
MDL Number: MFCD00026750
CAS Number: 14857-34-2
EINECS: 238-921-7
ADR: 3,II

Order Now

Specification Test Results

Assay 94%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR