Cyclohexanone ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Pimelic Ketone
215915-1L
215915-4L
215915-500ML
BT215915-1LEA
51.68
USD
BT215915-1L
BT215915-4L
BT215915-500ML
Cyclohexanone ≥99.0% ACS
Cyclohexanone
CAS: 108-94-1; EC No: 203-631-1; MDL No: MFCD00001625; RTECS: GW1050000
UN No: UN1915; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H10O; MW: 98.15
Melting Point: -47°; Boiling Point: 155°; Flash point: 44°C (111°F)
Density (g/mL): 0.947; Refractive Index: 1.450
- RTECS: GW1050000
- Refractive index: 1.450
- Liquid
Formula:
C₆H₁₀(=O) Boiling Pt: 155 °C Melting Pt: –47 °C Density: 0.947 g/mL Flash Pt: 44 °C (111 °F) |
MDL Number:
MFCD00001625 CAS Number: 108-94-1 EINECS: 203-631-1 UN: 1915 ADR: 3,III |
|
Specification Test Results
Assay | 99.0% min. |
Color (APHA) | ≤10 |
Residue after evaporation | ≤0.05% |
Cyclohexanol | ≤0.05% |
Free acid (as formic acid) | ≤0.01% |
Water | ≤0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...