Dodecylamine ≥98%
Supplier: BeanTown Chemical
CAS: 124-22-1; EC No: 204-690-6; MDL No: MFCD00008154; RTECS: JR6475000
UN No: UN3259; Haz Class: 8; Packing Group: II
Low Melting Solid; Linear Formula: CH3(CH2)11NH2; Molecular Formula: C12H25N ; MW: 185.36
Melting Point: 27-29°; Boiling Point: 247-249°; Flash point: 115°C (239°F)
Density (g/mL): 0.806
- RTECS: JR6475000
- Low Melting Solid
Formula:
H₃C(CH₂)₁₀CH₂NH₂ Boiling Pt: 247…249 °C Melting Pt: 27…29 °C Density: 0.806 g/mL Flash Pt: 115 °C (239 °F) |
MDL Number:
MFCD00008154 CAS Number: 124-22-1 EINECS: 204-690-6 UN: 3259 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...