Spermidine ≥99%
Supplier: BeanTown Chemical
Synonyms:
N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
215885-1G
215885-25G
215885-5G
BT215885-1GEA
37.48
USD
BT215885-1G
BT215885-25G
BT215885-5G
Spermidine ≥99%
Spermidine
CAS: 124-20-9; EC No: 204-689-0; MDL No: MFCD00008229; RTECS: EJ7000000
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: NH2(CH2)3NH(CH2)4NH2; Molecular Formula: C7H19N3; MW: 145.25
Melting Point: 22-25°; Boiling Point: 128-130°/14 mmHg; Flash point: 112°C (234°F)
Density (g/mL): 0.925; Refractive Index: 1.479
Air Sensitive , Hygroscopic
- RTECS: EJ7000000
- Refractive index: 1.479
- Air Sensitive, Hygroscopic
- Liquid
Formula:
C₇H₁₉N₃ MW: 145.25 g/mol Boiling Pt: 128…130 °C/14 mmHg Melting Pt: 22…25 °C Density: 0.925 g/mL Flash Pt: 112 °C (234 °F) |
MDL Number:
MFCD00008229 CAS Number: 124-20-9 EINECS: 204-689-0 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...