Tantalum pentafluoride 99,9
Supplier: BeanTown Chemical
215815-10G
215815-50G
BT215815-10GEA
142
USD
BT215815-10G
BT215815-50G
Tantalum pentafluoride 99,9
Tantalum pentafluoride
CAS: 7783-71-3; EC No: 232-022-3; MDL No: MFCD00042542
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Linear Formula: TaF5; MW: 275.94
Melting Point: 97°; Boiling Point: 230°
Density (g/mL): 4.74
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
F₅Ta MW: 275.94 Boiling Pt: 229 Melting Pt: 97 Density: 4.74 g/mL |
MDL Number:
MFCD00042542 CAS Number: 7783-71-3 EINECS: 232-022-3 UN: 3260 ADR: 8 II Merck Index: 12,09225 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...