Methyl acrylate ≥99% stabilized

Supplier: BeanTown Chemical

215790-100ML 215790-2.5L 215790-500ML
BT215790-100MLEA 21.52 USD
BT215790-100ML BT215790-2.5L BT215790-500ML
Methyl acrylate ≥99% stabilized
Methyl acrylate (monomer)

CAS: 96-33-3; EC No: 202-500-6; MDL No: MFCD00008627; RTECS: AT2800000 UN No: UN1919; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH2=CHCOOCH3; Molecular Formula: C4H6O2; MW: 86.09 Melting Point: -75°; Boiling Point: 79-81°; Flash point: -3°C (27°F) Density (g/mL): 0.956; Refractive Index: 1.402


  • RTECS: AT2800000
  • Refractive index: 1.402
  • Liquid

Formula: C₄H₆O₂
Boiling Pt: 79…81 °C
Melting Pt: –75 °C
Density: 0.956 g/mL
Flash Pt: –3 °C (27 °F)
MDL Number: MFCD00008627
CAS Number: 96-33-3
EINECS: 202-500-6
UN: 1919
ADR: 3,II

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR