Cobalt(II,III) oxide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
215780-100G
215780-25G
215780-500G
BT215780-100GEA
110.63
USD
BT215780-100G
BT215780-25G
BT215780-500G
Cobalt(II,III) oxide ≥99.5%
Cobalt(II,III) oxide
CAS: 1308-06-1; EC No: 215-157-2; MDL No: MFCD00010939; RTECS: GG2500000
UN No: UN3077; Haz Class: 9; Packing Group: III
-400 Mesh Powder; Molecular Formula: Co3O4; MW: 240.80
Melting Point: 895° (decomposes)
Density (g/mL): 6.11
- RTECS: GG2500000
- -400 Mesh Powder
Formula:
Co₃O₄ MW: 240.8 g/mol Melting Pt: 895 (dec.) Density: 6.11 g/mL |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 ADR: 9 III Merck Index: 13,02454 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...