Chromium ≥99.95% (trace metals basis), powder -200 mesh
Supplier: BeanTown Chemical
215765-100G
215765-25G
215765-500G
BT215765-100GEA
250
USD
BT215765-100G
BT215765-25G
BT215765-500G
Chromium ≥99.95% (trace metals basis), powder -200 mesh
Chromium
CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944; RTECS: GB4200000
UN No: UN3089; Haz Class: 4.1; Packing Group: III
Powder; Molecular Formula: Cr; MW: 52.00
Melting Point: 1857°; Boiling Point: 2672°
Density (g/mL): 7.14
- RTECS: GB4200000
- Powder
Formula:
Cr MW: 52 Boiling Pt: 2469 Melting Pt: 1840 Density: 7.14 g/mL |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 UN: 3089 ADR: 4.1 III Merck Index: 12,02288 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...