1-Bromopropane ≥99%
Supplier: BeanTown Chemical
Synonyms:
Solvent 38, n-Propyl bromide
215735-1L
215735-250ML
BT215735-1LEA
119.73
USD
BT215735-1L
BT215735-250ML
1-Bromopropane ≥99%
1-Bromopropane
CAS: 106-94-5; EC No: 203-445-0; MDL No: MFCD00000254; RTECS: VA6020000
UN No: UN2344; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: CH3CH2CH2Br; Molecular Formula: C3H7Br; MW: 122.99
Melting Point: -110°; Boiling Point: 71°; Flash point: 21°C (70°F)
Refractive Index: 1.434
Light Sensitive
- RTECS: VA6020000
- Refractive index: 1.434
- Light Sensitive
- Liquid
Formula:
BrH₂CCH₂CH₃ MW: 122.99 g/mol Boiling Pt: 71 °C Melting Pt: –110 °C Flash Pt: 21 °C (70 °F) |
MDL Number:
MFCD00000254 CAS Number: 106-94-5 EINECS: 203-445-0 UN: 2344 ADR: 3,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...