1-Hexanol ≥99%

Supplier: BeanTown Chemical

Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

215690-2.5L 215690-500ML
BT215690-2.5LEA 104.84 USD
BT215690-2.5L BT215690-500ML
1-Hexanol ≥99%
1-Hexanol

CAS: 111-27-3; EC No: 203-852-3; MDL No: MFCD00002982; RTECS: MQ4025000 UN No: UN2282; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3(CH2)5OH; Molecular Formula: C6H14O; MW: 102.17 Melting Point: -52°; Boiling Point: 156-157° Density (g/mL): 0.814; Refractive Index: 1.418


  • RTECS: MQ4025000
  • Refractive index: 1.418
  • Liquid

Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
Boiling Pt: 156…157 °C
Melting Pt: –52 °C
Density: 0.814 g/mL
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR