Pivalonitrile ≥98%
Supplier: BeanTown Chemical
CAS: 630-18-2; EC No: 211-133-0; MDL No: MFCD00001847
UN No: UN3273; Haz Class: 3 (6.1); Packing Group: II
Liquid; Linear Formula: (CH3)3CCN; Molecular Formula: C5H9N; MW: 83.13
Melting Point: 15-16°; Boiling Point: 105-106°; Flash point: 4°C (39°F)
Density (g/mL): 0.752; Refractive Index: 1.377
- Refractive index: 1.377
- Liquid
Formula:
C₅H₉N Boiling Pt: 105…106 °C Melting Pt: 15…16 °C Density: 0.752 g/mL Flash Pt: 4 °C (39 °F) |
MDL Number:
MFCD00001847 CAS Number: 630-18-2 EINECS: 211-133-0 ADR: 3 (6.1),II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...