Silver nitrite ≥99%
Supplier: BeanTown Chemical
Synonyms:
Argentous nitrite
215575-10G
215575-50G
BT215575-10GEA
80
USD
BT215575-10G
BT215575-50G
Silver nitrite ≥99%
Silver nitrite
CAS: 7783-99-5; EC No: 232-041-7; MDL No: MFCD00003413
UN No: UN2627; Haz Class: 5.1; Packing Group: II
Crystalline; Molecular Formula: AgNO2; MW: 153.88
Melting Point: 140° (decomposes)
Density (g/mL): 4.453
Light Sensitive
- Light Sensitive
- Crystalline
Formula:
AgNO₂ MW: 153.87 g/mol Melting Pt: 140 °C (dec.) Density: 4.453 g/mL |
MDL Number:
MFCD00003413 CAS Number: 7783-99-5 EINECS: 232-041-7 UN: 2627 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...