2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride ≥97%
Supplier: BeanTown Chemical
215565-1G
215565-5G
BT215565-1GEA
35
USD
BT215565-1G
BT215565-5G
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride ≥97%
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride
CAS: 72830-09-2; EC No: 416-440-5; MDL No: MFCD02181083
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C8H11Cl2NO2; MW: 224.08
Melting Point: 155° (decomposes)
- Solid
Formula:
C₈H₁₀ClNO₂ Melting Pt: 155 °C (dec.) |
MDL Number:
MFCD02181083 CAS Number: 72830-09-2 EINECS: 416-440-5 ADR: 9,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...