Butyric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: Butanoic acid

215395-100ML 215395-2.5L 215395-500ML
BT215395-100MLEA 27.85 USD
BT215395-100ML BT215395-2.5L BT215395-500ML
Butyric acid ≥99%
Butyric acid

CAS: 107-92-6; EC No: 203-532-3; MDL No: MFCD00002814; RTECS: ES5425000 UN No: UN2820; Haz Class: 8; Packing Group: III Liquid; Linear Formula: CH2CH2CH2COOH; Molecular Formula: C4H8O2; MW: 88.11 Melting Point: -6°; Boiling Point: 162°; Flash point: 72°C (162°F) Density (g/mL): 0.964; Refractive Index: 1.398


  • RTECS: ES5425000
  • Refractive index: 1.398
  • Liquid

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 162 °C
Melting Pt: –6 °C
Density: 0.964 g/mL
Flash Pt: 72 °C (162 °F)
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: 8,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR