Thymol ≥98%
Supplier: BeanTown Chemical
CAS: 89-83-8; EC No: 201-944-8; MDL No: MFCD00002309; RTECS: XP2275000
UN No: UN2430; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: [(CH3)2CH]C6H3(OH)(CH3); Molecular Formula: C10H14O; MW: 150.22
Melting Point: 48-51°; Boiling Point: 231-233°; Flash point: 102°C (215°F)
Density (g/mL): 0.965
- RTECS: XP2275000
- Crystalline
Formula:
2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH Boiling Pt: 231…233 °C Melting Pt: 48…51 °C Density: 0.965 g/mL Flash Pt: 102 °C (215 °F) |
MDL Number:
MFCD00002309 CAS Number: 89-83-8 EINECS: 201-944-8 UN: 2430 ADR: 8,III |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...