4-Iodobiphenyl ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Iodo-1,1'-biphenyl
215300-1G
215300-25G
215300-5G
BT215300-1GEA
41
USD
BT215300-1G
BT215300-25G
BT215300-5G
4-Iodobiphenyl ≥97%
4-Iodobiphenyl
CAS: 1591-31-7; EC No: 216-469-1; MDL No: MFCD00019028
UN No: UN3077; Haz Class: 9; Packing Group: III
Crystalline/Powder; Molecular Formula: C12H9I; MW: 280.10
Melting Point: 110-114°
Light Sensitive
- Light Sensitive
- Crystalline/Powder
Formula:
C₁₂H₉I Melting Pt: 110…114 °C |
MDL Number:
MFCD00019028 CAS Number: 1591-31-7 EINECS: 216-469-1 ADR: 9,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...