Potassium-2-ethylhexanoate solution 75% (w/w) ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Potassium 2-ethylhexanoate, 2-Ethylhexanoic Acid Potassium Salt
215265-25G
215265-5G
BT215265-25GEA
275
USD
BT215265-25G
BT215265-5G
Potassium-2-ethylhexanoate solution 75% (w/w) ≥99.9% (trace metals basis)
Potassium-2-ethylhexanoate
CAS: 3164-85-0; EC No: 221-625-7; MDL No: MFCD00045896
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)3CH(CH2CH3)COOK; Molecular Formula: C8H15KO2; MW: 182.30
Moisture Sensitive
- Moisture Sensitive
Formula:
C₈H₁₆O₂ MW: 182.30 g/mol |
MDL Number:
MFCD00045896 CAS Number: 3164-85-0 EINECS: 221-625-7 UN: 1993 ADR: 3 III |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...