Acrylamide ≥98%

Supplier: BeanTown Chemical

215245-100G 215245-2.5KG 215245-500G
BT215245-100GEA 24.23 USD
BT215245-100G BT215245-2.5KG BT215245-500G
Acrylamide ≥98%
Acrylamide

CAS: 79-06-1; EC No: 201-173-7; MDL No: MFCD00008032; RTECS: AS3325000 UN No: UN2074; Haz Class: 6.1; Packing Group: III Crystalline; Linear Formula: CH2=CHCONH2; Molecular Formula: C3H5NO; MW: 71.08 Melting Point: 82-86°; Boiling Point: 125°/25 mmHg; Flash point: 138°C (280°F) Light Sensitive


  • RTECS: AS3325000
  • Light Sensitive
  • Crystalline

Formula: C₃H₅NO
Boiling Pt: 125 °C/25 mmHg
Melting Pt: 82…86 °C
Flash Pt: 138 °C (280 °F)
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7687b7f17a8cdf31c559a33fd5d4021a2e50a40a&vid=fecebe0c34b037788df119c11bfc9637d78cda0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR