Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
215200-1G
215200-250MG
215200-5G
BT215200-1GEA
176
USD
BT215200-1G
BT215200-250MG
BT215200-5G
Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (trace metals basis)
Dihydrogen hexabromoosmate(IV) hydrate
MDL No: MFCD01863474
UN No: UN3260; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: H2OsBr6·xH2O; MW: 352.02 (anhydrous)
- Crystalline
Formula:
H₂OsBr₆·xH₂O MW: 352.02 g/mol (anhy) |
MDL Number:
MFCD01863474 ADR: 8,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...