3-Nitro-p-toluidine ≥95%
Supplier: BeanTown Chemical
Synonyms:
3-Nitro-4-methylaniline, 4-Methyl-3-nitroaniline, 4-Amino-2-nitrotoluene
215100-100G
215100-25G
215100-5G
BT215100-100GEA
246
USD
BT215100-100G
BT215100-25G
BT215100-5G
3-Nitro-p-toluidine ≥95%
3-Nitro-p-toluidine
CAS: 119-32-4; EC No: 201-924-9; MDL No: MFCD00007910; RTECS: XU8227250
UN No: UN2660; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: C7H8N2O2 ; MW: 152.15
Melting Point: 74-77°
- RTECS: XU8227250
- Powder
Formula:
C₇H₈N₂O₂ MW: 152.15 g/mol Melting Pt: 74…77 °C |
MDL Number:
MFCD00007910 CAS Number: 119-32-4 EINECS: 201-924-9 UN: 2660 ADR: 6.1,III |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...