Trizinc diarsenide ≥99.999%
Supplier: BeanTown Chemical
Synonyms:
Zinc arsenide, Trizinc diarsenide
215065-1G
215065-5G
BT215065-1GEA
55
USD
BT215065-1G
BT215065-5G
Trizinc diarsenide ≥99.999%
Trizinc diarsenide
CAS: 12006-40-5; EC No: 234-486-2; MDL No: MFCD00049629
UN No: UN1557; Haz Class: 6.1; Packing Group: II
Pieces; Linear Formula: Zn3As2; MW: 345.98
Melting Point: 1015°
Density (g/mL): 5.53
- Pieces
Formula:
As₂Zn₃ MW: 346.01 Melting Pt: 1015 °C Density: 5.53 g/mL |
MDL Number:
MFCD00049629 CAS Number: 12006-40-5 EINECS: 234-486-2 UN: 1557 ADR: 6.1 II |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...