N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA
215015-10G
215015-50G
BT215015-10GEA
43
USD
BT215015-10G
BT215015-50G
N,O-Bis(trimethylsilyl)trifluoroacetamide ≥98%
N,O-Bis(trimethylsilyl)trifluoroacetamide
CAS: 25561-30-2; EC No: 247-103-9; MDL No: MFCD00008269
UN No: UN2924; Haz Class: 3(8); Packing Group: III
Liquid; Linear Formula: CF3C[=NSi(CH3)3]OSi(CH3)3; Molecular Formula: C8H18F3NOSi2; MW: 257.40
Melting Point: -10°; Boiling Point: 145-147°; Flash point: -9°C (16°F)
Density (g/mL): 0.969; Refractive Index: 1.384
Moisture Sensitive
- Refractive index: 1.384
- Moisture Sensitive
- Liquid
Formula:
C₈H₁₈F₃NOSi₂ MW: 257.4 g/mol Boiling Pt: 145…147 °C Melting Pt: –10 °C Density: 0.969 g/mL Flash Pt: –9 °C (16 °F) |
MDL Number:
MFCD00008269 CAS Number: 25561-30-2 EINECS: 247-103-9 UN: 2924 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...