Acrylonitrile ≥99% stabilized

Supplier: BeanTown Chemical
Danger

215010-100ML 215010-2.5L 215010-500ML
BT215010-100MLEA 22 USD
BT215010-100ML BT215010-2.5L BT215010-500ML
Acrylonitrile ≥99% stabilized
Acrylonitrile

CAS: 107-13-1; EC No: 203-466-5; MDL No: MFCD00001927; RTECS: AT5250000 UN No: UN1093; Haz Class: 3 (6.1); Packing Group: I Liquid; Linear Formula: CH2=CHCN; MW: 53.06 Melting Point: -83°; Boiling Point: 77°; Flash point: -5°C (23°F) Refractive Index: 1.391 Light Sensitive


  • RTECS: AT5250000
  • Light Sensitive

Formula: C₃H₃N
MW: 53.06 g/mol
Boiling Pt: 77 °C
Melting Pt: –83 °C
MDL Number: MFCD00001927
CAS Number: 107-13-1
EINECS: 203-466-5
UN: 1093
ADR: 3,II
Merck Index: 14,131

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR