Praseodymium ≥99.9% (trace rare earth metals basis), ingot

Supplier: BeanTown Chemical

214980-10G 214980-50G
BT214980-10GEA 119.71 USD
BT214980-10G BT214980-50G
Praseodymium ≥99.9% (trace rare earth metals basis), ingot
Praseodymium

CAS: 7440-10-0; EC No: 231-120-3; MDL No: MFCD00011174 UN No: UN3178; Haz Class: 4.1; Packing Group: II Solid; Molecular Formula: Pr; MW: 140.91 Melting Point: 931°; Boiling Point: 3520° Density (g/mL): 6.71


  • Solid

Formula: Pr
MW: 140.91 g/mol
Boiling Pt: 3520 °C
Melting Pt: 931 °C
Density: 6.71 g/mL
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
ADR: 4.1,II

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR