Tantalum 99,95, powder

Supplier: BeanTown Chemical

214960-100G 214960-25G 214960-5G
BT214960-100GEA 402.9 USD
BT214960-100G BT214960-25G BT214960-5G
Tantalum 99,95, powder
Tantalum

CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Ta; MW: 180.95 Melting Point: 2996°; Boiling Point: 5425° Density (g/mL): 16.69


  • RTECS: WW5505000
  • Powder

Formula: Ta
MW: 180.95 g/mol
Boiling Pt: 5427
Melting Pt: 2996
Density: 16.69 g/mL
MDL Number: MFCD00011252
CAS Number: 7440-25-7
EINECS: 231-135-5
UN: 3089
ADR: 4.1 II
Merck Index: 12,09223

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR