Diphenylphosphine
Supplier: BeanTown Chemical
CAS: 829-85-6; EC No: 212-591-4; MDL No: MFCD00003040
UN No: UN2845; Haz Class: 4.2; Packing Group: I
Liquid; Linear Formula: (C6H5)2PH ; Molecular Formula: C12H11P; MW: 186.19
Density (g/mL): 1.07; Refractive Index: 1.625
Air Sensitive, Moisture Sensitive
- Refractive index: 1.625
- Air Sensitive, Moisture Sensitive
- Liquid
Formula:
C₁₂H₁₁P Density: 1.07 g/mL |
MDL Number:
MFCD00003040 CAS Number: 829-85-6 EINECS: 212-591-4 UN: 2845 ADR: 4.2,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...