Boron nitride ≥97% aerosol refractory paint
Supplier: BeanTown Chemical
214930-13OZ
214930-5X13OZ
BT214930-13OZEA
125
USD
BT214930-13OZ
BT214930-5X13
Boron nitride ≥97% aerosol refractory paint
Boron nitride
CAS: 10043-11-5; MDL No: MFCD00011317
UN No: UN1950; Haz Class: 2.1
Aerosol Spray Can; Molecular Formula: BN; MW: 24.82
- Aerosol Spray Can
Formula:
BN MW: 24.82 g/mol |
MDL Number:
MFCD00011317 CAS Number: 10043-11-5 ADR: 2.1, |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...