2,4,6-Collidine ≥99%
Supplier: BeanTown Chemical
CAS: 108-75-8; EC No: 203-613-3; MDL No: MFCD00006338; RTECS: UU0970000
UN No: UN1992; Haz Class: 3 (6.1); Packing Group: III
Liquid; Molecular Formula: C8H11N; MW: 121.18
Melting Point: -43°; Boiling Point: 171-172°; Flash point: 55°C (131°F)
Density (g/mL): 0.917; Refractive Index: 1.498
- RTECS: UU0970000
- Refractive index: 1.498
- Liquid
Formula:
C₈H₁₁N Boiling Pt: 171…172 °C Melting Pt: –43 °C Density: 0.917 g/mL Flash Pt: 55 °C (131 °F) |
MDL Number:
MFCD00006338 CAS Number: 108-75-8 EINECS: 203-613-3 UN: 1992 ADR: 3,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...